missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Phospholamban (Ser16) Recombinant Rabbit Monoclonal Antibody (K01_1W72)

Product Code. p-200114709
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826187 100 μL
30826717 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826187 Supplier Invitrogen™ Supplier No. MA563575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Phospho-Phospholamban (Ser16) Recombinant Monoclonal Antibody for Western Blot

Phospholamban (PLB) is a 52 amino acid phosphoprotein which regulates the calcium pump of cardiac sarcoplasmic reticulum (SR). PLB is an oligomer of five identical subunits each having a cytoplasmic and transmembrane domain. The cytoplasmic domain (residues 1 to 25) contains the phosphorylation sites and is highly basic and readily cleaved by proteases whereas the transmembrane domain (residues 25 to 52) is mostly hydrophobic, protease resistant and stabilizes the pentamer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Phospholamban (Ser16)
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1W72
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene PLN
Gene Accession No. P26678, P61014
Gene Alias cardiac CMH18; Cardiac phospholamban; CMD1P; CMH18; phospholamban; Plb; Plm; PLN; unnamed protein product
Gene Symbols PLN
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Ser16 of human Phospholamban.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18821, 5350
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.