missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PKC alpha (Thr497) Recombinant Rabbit Monoclonal Antibody (23GB4955), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | Phospho-PKC alpha (Thr497) |
|---|---|
| Clone | 23GB4955 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30089631
|
Invitrogen™
MA553806 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088060
|
Invitrogen™
MA553807 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-PKC alpha (Thr497) Recombinant Monoclonal antibody specifically detects Phospho-PKC alpha (Thr497) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| Phospho-PKC alpha (Thr497) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P05132, P05696, P17252, P20444, P27791 | |
| 18747, 18750, 24680, 25636, 5566 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB4955 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| AAG6; aging-associated gene 6; AI875142; EC 2.7.11.13; FHC; FTH; FTHL6; im:7139045; kinase PKC-alpha; KPCA; MGC104426; PIG15; PKC III; Pkca; PKC-A; PKC-alpha; PKC-III; PKRCA; PLIF; PRKACA; Prkca; prkcaa; protein kinase C; protein kinase C alpha; protein kinase C alpha type; protein kinase C alpha type; protein kinase C, alpha, a; protein kinase C, alpha; protein kinase C, alpha, a; protein kinase c-alpha; si:dkey-8l19.1; Unknown (protein for IMAGE:7139045); unnamed protein product | |
| A synthesized peptide derived from human Phospho-PKC alpha (Thr497) (490-497AA). | |
| Primary | |
| -20°C | |
| PRKACA |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title