missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PKC zeta (Thr410) Polyclonal Antibody

Description
Positive Control: HeLa Predicted Reactivity: Mouse (100%), Rat (100%), Zebrafish (100%), Pig (100%), Rabbit (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
Specifications
Specifications
| Antigen | Phospho-PKC zeta (Thr410) |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.81 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol, 1% BSA and 0.025% ProClin 300; pH 7 |
| Gene | PRKCZ |
| Gene Accession No. | Q05513 |
| Gene Alias | 14 - 3 - 3 - zeta isoform; 14-3-3-zetaisoform; AI098070; aPKCzeta; atypical protein kinase C; C80388; kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; PKC2; Pkcz; PKC-ZETA; Prkcz; protein kinase C zeta; protein kinase C zeta subspecies; protein kinase C zeta type; protein kinase C, zeta; r14-3-3; R74924; zetaPKC |
| Gene Symbols | PRKCZ |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?