missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-PKR (Thr258) Polyclonal Antibody

Phospho-PKR (Thr258) Polyclonal Antibody
Brand: Invitrogen PA539751
This item is not returnable.
View return policy
Description
Phospho-PKR (Thr258) Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| Phospho-PKR (Thr258) | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK2 | |
| 2310047A08Rik; 4732414G15Rik; AI467567; AI747578; double stranded RNA activated protein kinase; dsRNA-activated kinase; EC 2.7.11.1; eIF-2 alpha; eIF2a Kinase; eIF-2A protein kinase 2; EIF2AK1; EIF2AK2; eukaryotic translation initiation factor 2 alpha kinase 2; eukaryotic translation initiation factor 2-alpha kinase 2; IFN- type I-induced and dsRNA-activated kinase; IFN-induced and double-stranded RNA-activated kinase; interferon-induced, double-stranded RNA-activated protein kinase; interferon-inducible elF2alpha kinase; interferon-inducible RNA-dependent protein kinase; MGC126524; OTTHUMP00000201320; P1/eIF-2A protein kinase; p68 kinase; PKR; PPP1R83; Prkr; protein kinase R; protein kinase RNA-activated; protein kinase, interferon inducible double stranded RNA dependent; Protein kinase, interferon-inducible double stranded RNA dependent; protein phosphatase 1, regulatory subunit 83; serine/threonine-protein kinase TIK; T-cell viral integration site; Tik; Tyrosine-protein kinase EIF2AK2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5610 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19525 | |
| EIF2AK2 | |
| A synthetic phosphopeptide derived from human PKR around the phosphorylation site of Thr258 (K-Y-TP-V-D). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction