missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-PRAS40 (Thr246) Recombinant Rabbit Monoclonal Antibody (23GB3855), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 217.00 - € 590.00
Specifications
| Antigen | Phospho-PRAS40 (Thr246) |
|---|---|
| Clone | 23GB3855 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089316
![]() |
Invitrogen™
MA553652 |
100 μL |
€ 590.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087685
|
Invitrogen™
MA553653 |
20 μL |
€ 217.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-PRAS40 (Thr246) Recombinant Monoclonal antibody specifically detects Phospho-PRAS40 (Thr246) in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| Phospho-PRAS40 (Thr246) | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q96B36, Q9D1F4 | |
| 292887, 67605, 84335 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB3855 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 1110012J22Rik; 40 kDa proline-rich AKT substrate; AI227026; AKT1 substrate 1; AKT1 substrate 1 (proline rich); AKT1 substrate 1 (proline-rich); akt1s1; Lobe; Lobel; OTTHUMP00000196756; OTTHUMP00000196760; PKB/Akt substrate 40; Pras; PRAS40; Proline-rich AKT substrate; proline-rich AKT1 substrate 1 | |
| A synthetic peptide of human Phospho-PRAS40 (Thr246) (239-253AA). | |
| Primary | |
| -20°C | |
| AKT1S1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title