missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PRK1/PRK2 (Thr774, Thr816) Polyclonal Antibody
GREENER_CHOICE

Product Code. 11362932
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362932 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11362932 Supplier Invitrogen™ Supplier No. PA517823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

Receptor-like kinase involved in the control of pollen germination and pollen tube polar growth.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PRK1/PRK2 (Thr774, Thr816)
Applications Western Blot
Classification Polyclonal
Concentration 42 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PKN1
Gene Accession No. O08874, P70268, Q16512, Q16513, Q63433, Q8BWW9
Gene Alias 6030436C20Rik; AI507382; cardiolipin-activated protein kinase Pak2; DBK; F730027O18Rik; p140 kinase; PAK1; PAK-1; PAK2; Pak-2; Pkn; PKN gamma; Pkn1; Pkn2; PKN-ALPHA; Prk1; Prk2; Prkcl1; PRKCL2; PRO2042; protease-activated kinase 1; protease-activated kinase 2; protein kinase C-like 1; Protein kinase C-like 2; protein kinase C-like PKN; protein kinase C-related kinase 1; protein kinase N1; protein kinase N2; protein kinase PKN; protein kinase PKN-alpha; Protein-kinase C-related kinase 1; protein-kinase C-related kinase 2; serine/threonine kinase 3; serine/threonine kinase 7; serine/threonine protein kinase N; serine/threonine-protein kinase N1; Serine/threonine-protein kinase N2; serine-threonine kinase N; serine-threonine protein kinase N; Stk3; Stk7
Gene Symbols PKN1, PKN2
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding pThr774 of human PRK1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 109333, 207122, 29355, 320795, 5585, 5586
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.