missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-PTRF (Tyr156) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16643825
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16643825 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16643825 Supplier Invitrogen™ Supplier No. PA599575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of PTRF only when phosphorylated at Tyr156.

The polymerase I and transcript release factor (PTRF) enables the dissociation of paused ternary polymerase I transcription complexes from the 3' end of pre-rRNA transcripts, regulating rRNA transcription by promoting the dissociation of transcription complexes and the re-initiation of polymerase I on nascent rRNA transcripts. PTRF also localizes to caveolae at the plasma membrane and is thought to play a critical role in the formation of caveolae and the stabilization of caveolins, translocating from caveolae to the cytoplasm after insulin stimulation. PTRF is also thought to modify lipid metabolism and insulin-regulated gene expression. Mutations in this gene result in a disorder characterized by generalized lipodystrophy and muscular dystrophy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-PTRF (Tyr156)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CAVIN1
Gene Accession No. O54724, P85125, Q6NZI2
Gene Alias 2310075E07Rik; AW546441; caveolae associated protein 1; caveolae-associated protein 1; CAVIN; CAVIN1; cavin-1; Cav-p60; CGL4; FKSG13; FLJ90031; Polymerase I and transcript release factor; PTRF; ptrf {ECO:0000312; RGD:1307685}; RNA polymerase I and transcript release factor; TTF-I interacting peptide 12
Gene Symbols CAVIN1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CAVIN1(Accession Q6NZI2), corresponding to amino acid residues around phosphorylated Tyr156.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19285, 284119, 287710
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.