missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RIP1 (Ser166) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA5117250
This item is not returnable.
View return policy
Description
Positive Samples: NIH/3T3
Phospho-RIP1 (Ser166) Polyclonal antibody specifically detects Phospho-RIP1 (Ser166) in Human, Mouse, Mouse samples. It is validated for Western Blot
Specifications
| Phospho-RIP1 (Ser166) | |
| Polyclonal | |
| Unconjugated | |
| RIPK1 | |
| Cell death protein RIP; D330015H01Rik; FLJ39204; OTTHUMP00000015955; receptor (TNFRSF)-interacting serine-threonine kinase 1; receptor interacting serine/threonine kinase 1; receptor-interacting protein 1; receptor-interacting protein kinase 1; receptor-interacting serine/threonine-protein kinase 1; Rinp; RIP; RIP1; RIP-1; RIP1 (CT); RIPK1; serine/threonine-protein kinase RIP | |
| A phospho specific peptide corresponding to residues surrounding S166 of Mouse RIPK1 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| 3.01 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q13546, Q60855 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 19766, 8737 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction