missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RPA2 (Ser4, Ser8) Recombinant Rabbit Monoclonal Antibody (BL-165-5F1)

Rabbit Recombinant Monoclonal Antibody
€ 148.00 - € 376.00
Specifications
| Antigen | Phospho-RPA2 (Ser4, Ser8) |
|---|---|
| Clone | BL-165-5F1 |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17795579
![]() |
Invitrogen™
MA544213 |
100 μL |
€ 376.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17705589
|
Invitrogen™
MA544214 |
20 μL |
€ 148.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phospho-RPA2 (Ser4, Ser8) Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IP
Required for DNA recombination, repair and replication. The activity of RP-A is mediated by single-stranded DNA binding and protein interactions. Required for the efficient recruitment of the DNA double-strand break repair factor RAD51 to chromatin in response to DNA damage.Specifications
| Phospho-RPA2 (Ser4, Ser8) | |
| 1 mg/mL | |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| BBS with 0.09% sodium azide; pH 8.2 | |
| P15927 | |
| 6118 | |
| Dually phosphorylated synthetic peptide, which represented a portion of human replication protein A2, 32 kDa surrounding phosphorylated serines that corresponded to positions 4 and 8. | |
| Antibody |
| BL-165-5F1 | |
| 4°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Rpa2 | |
| 30-kDa protein; AA409079; AI325195; AU020965; HSSB; hypothetical protein; I79_004200; p32; p32-subunit of replication protein A; p34; REPA2; Replication factor A protein 2; replication protein A 32 kDa subunit; Replication protein A 32kDa subunit; Replication protein A 34 kDa subunit; Replication protein A 34kDa subunit; replication protein A2; replication protein A2, 32kDa; replication protein A4; RFA; RF-A protein 2; Rf-A2; RP21C; RP4-547C9.3; RP-A p32; RP-A p34; RPA2; RPA32; RPA34; RPA4; RpLP1; RpP2 | |
| Rpa2 | |
| Primary | |
| purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title