missing translation for 'onlineSavingsMsg'
Learn More

Phospho-RPS6KA1 (Thr359/Ser363) Rabbit Recombinant Antibody, Proteintech

Product Code. p-200108290 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30236164 20 μL 20µL
30234376 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30236164 Supplier Proteintech Supplier No. 814751RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The Phospho-RPS6KA1 (Thr359/Ser363) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-RPS6KA1 (Thr359/Ser363). This antibody recognizes human, mouse antigen. The Phospho-RPS6KA1 (Thr359/Ser363) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.

Specifications

Antigen RPS6KA1 (Thr359/Ser363)
Applications Western Blot, Flow Cytometry (Intracellular Staining), ELISA
Classification Recombinant Monoclonal
Clone 3O10
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols HU-1; MAPKAPK1A; RSK; RSK1
Host Species Rabbit
Immunogen Peptide
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6195
Target Species Human, Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 83-90 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.