missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-RyR3 (Ser160) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105062
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of RYR3 only when phosphorylated at Ser160.
RYR3 is a ryanodine receptor, which functions to release calcium from intracellular storage for use in many cellular processes. RYR3 is involved in skeletal muscle contraction by releasing calcium from the sarcoplasmic reticulum followed by depolarization of T-tubules. Two transcript variants encoding different isoforms of RYR3 have been found. RYR3 mediates calcium channel release of Ca(2+) from the sarcoplasmic reticulum into the cytoplasm in muscle and plays a role in triggering muscle contraction. Further, RYR3 may also regulate Ca(2+) release by other calcium channels, as well as Ca(2+) release from the endoplasmic reticulum in non-muscle cells. RYR3 also contributes to cellular calcium ion homeostasis. Plays a role in cellular calcium signaling. Diseases associated with RYR3 include Central Core Disease and Arrhythmogenic Right Ventricular Dysplasia 2.
Specifications
| Phospho-RyR3 (Ser160) | |
| Polyclonal | |
| Unconjugated | |
| RYR3 | |
| AI851294; Brain ryanodine receptor-calcium release channel; brain-type ryanodine receptor; C230090H21; calcium release channel; HBRR; Ryanodine receptor 3; RYR3; RYR-3; Type 3 ryanodine receptor | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 20192, 6263 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AGL3, Q15413 | |
| RYR3 | |
| A synthesized peptide derived from human RYR3(Accession Q15413), corresponding to amino acid residues around phosphorylated Ser160. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction