missing translation for 'onlineSavingsMsg'
Learn More

Phospho-S6 Ribosomal protein (Ser235) Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

€ 135.00 - € 360.00

Specifications

Antigen S6 Ribosomal protein (Ser235)
Clone 2A4B6
Concentration 500 μg/mL
Applications Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30234450
View Documents
Proteintech
67898-1-IG-20UL
20 μL
€ 135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30234565
View Documents
Proteintech
67898-1-IG-100UL
100 μL
€ 360.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Phospho-S6 Ribosomal protein (Ser235) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-S6 Ribosomal protein (Ser235). This antibody recognizes human, mouse, rat antigen. The Phospho-S6 Ribosomal protein (Ser235) antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.
Specifications

Specifications

S6 Ribosomal protein (Ser235)
500 μg/mL
Monoclonal
Liquid
RUO
6194
IgG2b
Protein A purification
Monoclonal Antibody
2A4B6
Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA
Unconjugated
Mouse
Human, Mouse, Rat
Peptide
Primary
Store at -20°C. Stable for one year after shipment.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.