missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-S6 Ribosomal protein (Ser240/244) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
€ 129.84 - € 401.60
Specifications
| Antigen | S6 Ribosomal protein (Ser240/244) |
|---|---|
| Clone | 5J13 |
| Concentration | 400 μg/mL |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Recombinant Monoclonal |
Description
The Phospho-S6 Ribosomal protein (Ser240/244) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-S6 Ribosomal protein (Ser240/244). This antibody recognizes human antigen. The Phospho-S6 Ribosomal protein (Ser240/244) antibody has been validated for the following applications: WB, IF/ICC, FC (Intra), ELISA analysis.Specifications
| S6 Ribosomal protein (Ser240/244) | |
| 400 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 6194 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
| 5J13 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title