missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SHC3 (Tyr424) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564710
This item is not returnable.
View return policy
Description
Phospho-SHC3 (Tyr424) Polyclonal Antibody detects endogenous levels of SHC3 only when phosphorylated at Tyr424.
Signaling adapter that couples activated growth factor receptors to signaling pathway in neurons. Involved in the signal transduction pathways of neurotrophin-activated Trk receptors in cortical neurons.
Specifications
| Phospho-SHC3 (Tyr424) | |
| Polyclonal | |
| Unconjugated | |
| SHC3 | |
| neuronal Shc; Nshc; N-Shc; Protein Rai; RAI; SH2 domain protein C3; SHC (Src homology 2 domain containing) transforming protein 3; SHC adaptor protein 3; SHC3; Shc3 p51; SHCC; SHC-transforming protein 3; SHC-transforming protein C; src homology 2 domain-containing transforming protein C3; src homology 2 domain-containing-transforming protein C3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114858, 20418, 53358 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O70143, Q61120, Q92529 | |
| SHC3 | |
| A synthesized peptide derived from human Shc3 (Phospho-Tyr424). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction