missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SHIP1 (Tyr865) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105684
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SHIP1 only when phosphorylated at Tyr865.
This gene is a member of the inositol polyphosphate-5-phosphatase (INPP5) family and encodes a protein with an N-terminal SH2 domain, an inositol phosphatase domain, and two C-terminal protein interaction domains. Expression of this protein is restricted to hematopoietic cells where its movement from the cytosol to the plasma membrane is mediated by tyrosine phosphorylation. At the plasma membrane, the protein hydrolyzes the 5' phosphate from phosphatidylinositol (3,4,5)-trisphosphate and inositol-1,3,4,5-tetrakisphosphate, thereby affecting multiple signaling pathways. Overall, the protein functions as a negative regulator of myeliod cell proliferation and survival. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| Phospho-SHIP1 (Tyr865) | |
| Polyclonal | |
| Unconjugated | |
| INPP5D | |
| 7a33; hp51CN; Inositol polyphosphate-5-phosphatase 145 kDa; inositol polyphosphate-5-phosphatase D; Inositol polyphosphate-5-phosphatase of 145 kDa; inositol polyphosphate-5-phosphatase, 145 kDa; inositol polyphosphate-5-phosphatase, 145kD; inositol polyphosphate-5-phosphatase, 145kDa; Inpp5d; MGC104855; MGC142140; MGC142142; OTTHUMP00000165069; OTTHUMP00000165070; OTTHUMP00000165071; OTTHUMP00000165072; OTTHUMP00000203442; p150Ship; phosphatidylin; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 1; Phosphatidylinositol 4,5-bisphosphate 5-phosphatase; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 1; Phosphatidylinositol-4,5-bisphosphate 5-phosphatase; SH2 domain-containing inositol 5'-phosphatase 1; SH2 domain-containing inositol phosphatase 1; SH2 domain-containing inositol-5'-phosphatase 1; SH2-containing inositol phosphatase SHIP; SHIP; SHIP1; SHIP-1; signaling inositol polyphosphate 5 phosphatase SIP-145; signaling inositol polyphosphate phosphatase SHIP II; SIP-145; Src homology 2 domain-containing inositol-5-phosphatase; s-SHIP | |
| Rabbit | |
| sequential chromatography | |
| RUO | |
| 3635 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q92835 | |
| INPP5D | |
| A synthesized peptide derived from human INPP5D(Accession Q92835), corresponding to amino acid residues around phosphorylated Tyr865. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction