Learn More
Invitrogen™ Phospho-Ship2 (Tyr1135) Recombinant Rabbit Monoclonal Antibody (Ship2Y1135-1D2), APC
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA528014
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
The protein encoded by this gene is an SH2-containing 5'-inositol phosphatase that is involved in the regulation of insulin function. The encoded protein also plays a role in the regulation of epidermal growth factor receptor turnover and actin remodeling. Additionally, this gene supports metastatic growth in breast cancer and is a valuable biomarker for breast cancer.
Specifications
| Phospho-Ship2 (Tyr1135) | |
| Recombinant Monoclonal | |
| APC | |
| INPPL1 | |
| 51C; 51C protein; ablSH3-binding protein; inositol polyphosphate phosphatase like 1; inositol polyphosphate phosphatase-like 1; inositol polyphosphate phosphatase-like protein 1; Inppl1; INPPL-1; OPSMD; Phosphatidylinositol 3,4,5-trisphosphate 5-phosphatase 2; phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase 2; protein 51C; SH2 domain-containing inositol 5'-phosphatase 2; SH2 domain-containing inositol phosphatase 2; SH2 domain-containing inositol-5'-phosphatase 2; SH2-containing inositol phosphatase 2; SHIP2; SHIP-2 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 16332, 3636 | |
| 4°C | |
| Liquid |
| Flow Cytometry | |
| Ship2Y1135-1D2 | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| O15357, Q6P549 | |
| INPPL1 | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr1135 of human phospho Ship2. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.