missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SLC22A8 (Ser4) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105324
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SLC22A8 only when phosphorylated at Ser4.
SLC22A8 is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. SLC22A8 is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney.The protein encoded by this gene is involved in the sodium-independent transport and excretion of organic anions, some of which are potentially toxic. The encoded protein is an integral membrane protein and appears to be localized to the basolateral membrane of the kidney. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Specifications
| Phospho-SLC22A8 (Ser4) | |
| Polyclonal | |
| Unconjugated | |
| SLC22A8 | |
| hOAT3; MGC24086; mOat3; OAT3; OCT3; organic anion transporter 3; rbOAT3; reduced in osteosclerosis transporter; renal organic anion transporter 3; rOat3; Roct; Slc22a8; solute carrier family 22 (organic anion transporter), member 8; solute carrier family 22 ,member 8; solute carrier family 22 member 8; solute carrier family 22, member 8 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 19879, 83500, 9376 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O88909, Q8TCC7, Q9R1U7 | |
| SLC22A8 | |
| A synthesized peptide derived from human SLC22A8(Accession Q8TCC7), corresponding to amino acid residues around phosphorylated Ser4. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction