missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-SMAD1 (Ser214) Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81433-1-RR-100UL
This item is not returnable.
View return policy
Description
The Phospho-SMAD1 (Ser214) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-SMAD1 (Ser214). This antibody recognizes human, mouse antigen. The Phospho-SMAD1 (Ser214) antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SMAD1 (Ser214) | |
| Recombinant Monoclonal | |
| 800 μg/mL | |
| BSP1; JV4-1; JV41; MADH1; MADR1 | |
| Peptide | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| 52 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3K8 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 4086 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction