Learn More
Invitrogen™ Phospho-SMAD3 (Ser423, Ser425) Monoclonal Antibody (S.434.0)
Rabbit Monoclonal Antibody
Brand:  Invitrogen™ MA514936
Product Code. 11584901
  • € 536.00 / 100µL
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with other family members.
SMAD3 belongs to the SMAD family of proteins. They are similar to the gene products of the Drosophila gene 'mothers against decapentaplegic' (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein functions as a transcriptional modulator activated by transforming growth factor-beta and is thought to play a role in the regulation of carcinogenesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phospho-SMAD3 (Ser423, Ser425)
Monoclonal
1387 μg/mL
0.01M HEPES with 0.15M NaCl, 50% glycerol, 100 μg/mL BSA and <0.02% sodium azide; pH 7.5
P84022, P84025, Q8BUN5
Smad3
Synthetic phosphopeptide corresponding to residues surrounding pSer423/425 of Smad3.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ChIP Assay, Immunoprecipitation, Western Blot
S.434.0
Unconjugated
Smad3
AU022421; DKFZp586N0721; DKFZp686J10186; hMAD-3; hSMAD3; HSPC193; HsT17436; JV15-2; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 3; MAD homolog 3a; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MAD-3; madh3; madh3a; madh3-A; MGC60396; mMad3; mothers against decapentaplegic homolog 3; mothers against decapentaplegic homolog 3a; mothers against DPP homolog 3; SMA- and MAD-related protein 3; SMAD; Smad 3; SMAD family member 3; SMAD family member 3 L homeolog; SMAD family member 3a; SMAD, mothers against DPP homolog 3; Smad3; smad3.L; smad3a; TGF beta response effector Smad3; TGF-beta response effector Smad3; transcription factor; transcription factor Smad3; wu:fa99e03; XELAEV_18018454mg; XenMLP; Xmad3; XSmad3
Rabbit
Affinity chromatography
RUO
17127, 25631, 4088
-20°C