missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Smad5 (Ser463,Ser465) Recombinant Rabbit Monoclonal Antibody (8F4B10)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535867
This item is not returnable.
View return policy
Description
Immunogen sequence: SSVS.
SMADs are members of the MAD-related family of molecules. MAD-related proteins are a family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily. SMADs can be divided into receptor-regulated SMADs (R-SMADs: SMAD1, 2, 5, 8 and 9), common-mediator SMAD (co-SMAD: SMAD4), and inhibitory SMADs (I-SMADs: SMAD6 and 7). SMAD1, 5, 8 and 9 have high degrees of homology and antibodies are available that recognize sequences common to all of them. SMAD8 and SMAD9 are typically used as alternate names for one another in the literature.
Specifications
| Phospho-Smad5 (Ser463,Ser465) | |
| Recombinant Monoclonal | |
| 0.34 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99717 | |
| SMAD5 | |
| A synthetic phosphorylated peptide around S463 & S465 of human Smad5 (Q99717). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8F4B10 | |
| Unconjugated | |
| SMAD5 | |
| 1110051M15Rik; AI451355; DKFZp781C1895; DKFZp781O1323; dwarfin-C; DWFC; Dwf-C; hSmad5; JV5-1; MAD homolog 5; MAD, mothers against decapentaplegic homolog 5; Madh5; Mothers against decapentaplegic homolog 5; mothers against decapentaplegic, drosophila, homolog of, 5; mothers against DPP homolog 5; mSmad5; MusMLP; OTTHUMP00000223331; SMA- and MAD-related protein 5; SMAD; SMAD 5; SMAD family member 5; SMAD, mothers against DPP homolog 5; SMAD5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4090 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction