missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-SRF (Ser103) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104791
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of SRF only when phosphorylated at Ser103, which site historically referenced as Ser99.
Serum response factor (SRF) is a transcription factor that binds the serum response element (SRE), a sequence that mediates the transient response of many cellular genes to growth stimulation. SRF-binding sites are also constitutive promotor elements in many muscle-specific promotors. SFR is required for cardiac differentiation and maturation.
Specifications
| Phospho-SRF (Ser103) | |
| Polyclonal | |
| Unconjugated | |
| SRF | |
| AW049942; AW240594; c-fos serum response element-binding transcription factor; MCM1; minichromosome maintenance 1 homolog; OTTHUMP00000039820; RGD1559787; Serum response factor; serum response factor (c-fos serum response element-binding transcription factor); Srf | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 20807, 501099, 6722 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P11831, Q9JM73 | |
| SRF | |
| A synthesized peptide derived from human SRF(Accession P11831), corresponding to amino acid residues around phosphorylated Ser103. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction