Learn More
Phospho-STAT3 (Y705) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand:  R&D Systems IC46071T-100UG
Product Code. 18163751
  • € 541.00 / 100µg
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
STAT3
Monoclonal
Alexa Fluor 594
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor)
Phosphopeptide containing the human STAT3 Y705 site, Accession # P40763
100 μg
Primary
Reconstitution default not known
Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied.
IgG2b
Flow Cytometry
1004G
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
P40763
Rabbit
Protein A or G purified from cell culture supernatant
RUO
6774