Learn More
Phospho-STAT3 (Y705) Alexa Fluor™ 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems IC46071T-100UG
This item is not returnable.
View return policy
Description
STAT3 Monoclonal antibody specifically detects STAT3 in Human samples. It is validated for Flow Cytometry
Specifications
| STAT3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Acute-phase response factor, APRFMGC16063, DNA-binding protein APRF, FLJ20882, HIES, signal transducer and activator of transcription 3, signal transducer and activator of transcription 3 (acute-phase responsefactor) | |
| Phosphopeptide containing the human STAT3 Y705 site, Accession # P40763 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 1004G | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P40763 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 6774 | |
| Human | |
| Purified |