missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
€ 135.00 - € 360.00
Specifications
| Antigen | STAT5A (Tyr694)/STAT5B (Tyr699) |
|---|---|
| Clone | 1F7G5 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Flow Cytometry (Intracellular Staining), ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237050
|
Proteintech
68000-1-IG-20UL |
20 μL |
€ 135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234012
|
Proteintech
68000-1-IG-100UL |
100 μL |
€ 360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) antibody from Proteintech is a mouse monoclonal antibody generated with peptide of human Phospho-STAT5A (Tyr694)/STAT5B (Tyr699). This antibody recognizes human antigen. The Phospho-STAT5A (Tyr694)/STAT5B (Tyr699) antibody has been validated for the following applications: WB, FC (Intra), ELISA analysis.Specifications
| STAT5A (Tyr694)/STAT5B (Tyr699) | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| 6776 | |
| Peptide | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 1F7G5 | |
| Western Blot, Flow Cytometry (Intracellular Staining), ELISA | |
| Unconjugated | |
| Mouse | |
| Human | |
| MGF; STAT5 | |
| IgG2a | |
| Protein A purification | |
| Monoclonal Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title