missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-STAT6 (Y641) Alexa Fluor 750-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems IC3717S-100UG
This item is not returnable.
View return policy
Description
STAT6 Monoclonal specifically detects STAT6 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| STAT6 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
| Phosphopeptide containing human STAT6 Y641 site., Accession No. P42226 | |
| 100 μg | |
| 6778.0 | |
| Human | |
| Supernatant |
| Flow Cytometry | |
| 1248D | |
| Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
| P42226 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human STAT6 when phosphorylated at Y641. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction