missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Synapsin I (S603) Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems PPS036
This item is not returnable.
View return policy
Description
Prepared from rabbit serum by affinity purification via sequential chromatography on phospho- and dephosphopeptide affinity columns.
Synapsin I Polyclonal specifically detects Synapsin I in Human, Mouse, Rat, Bovine, Canine, Chicken, Zebrafish samples. It is validated for Western Blot.
Specifications
| Synapsin I (p ser603) | |
| Polyclonal | |
| Western Blot 1:1000 dilution | |
| SYN1 | |
| Phosphopeptide corresponding to amino acid residues surrounding the phospho-S603 of Synapsin I | |
| 100 μL | |
| Primary | |
| This antibody is specific for the 78 kDa Synapsin I phosphorylated at S603 in Western blots of rat brain lysates. | |
| For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
| Western Blot | |
| Unconjugated | |
| SYN1, SYN1a, SYN1b, Synapsin 1, synapsin IBrain protein 4.1, synapsin-1, SYNI | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6853 | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction