missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Talin (Ser425) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17816798
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17816798 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17816798 Supplier Invitrogen™ Supplier No. PA5143776

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects a 240 kDa* protein corresponding to the molecular mass of Talin on SDS-PAGE immunoblots of human A431, rat PC12, and rabbit fibroblast cells treated with Calyculin A, but is not observed after lambda phosphatase treatment.

Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Talin (Ser425)
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene TLN2
Gene Accession No. P26039, Q71LX4, Q9Y490, Q9Y4G6
Gene Alias 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; KIAA1027; mKIAA0320; RGD1565416; talin 1; talin 2; talin-1; Talin-2; TLN; Tln1; Tln2
Gene Symbols TLN1, TLN2
Host Species Rabbit
Immunogen Phospho-Talin (Ser-425) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding Ser-425 in human Talin 1. This sequence is conserved in rat, mouse, and chicken Talin 1, as well as in Talin 2 (Ser-428).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21894, 313494, 315776, 70549, 7094, 83660
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.