missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-TBC1D1 (Ser627) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105667
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of TBC1D1 only when phosphorylated at Ser627.
The product of this gene is a microtubule-associated protein that is predominantly expressed in cells of epithelial origin. Microtubule-associated proteins are thought to be involved in microtubule dynamics, which is essential for cell polarization and differentiation. This protein has been shown to be able to stabilize microtubules, and may serve to modulate microtubule functions. Studies of the related mouse protein also suggested an essential role in microtubule function required for spermatogenesis.
Specifications
| Phospho-TBC1D1 (Ser627) | |
| Polyclonal | |
| Unconjugated | |
| TBC1D1 | |
| 1110062G02Rik; AI385682; AW555803; Kiaa1108; LYN; Lyncein; mKIAA1108; TBC; Tbc1; TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1; TBC1 domain family member 1; TBC1 domain family, member 1; Tbc1d1; tre-2/USP6, BUB2, cdc16) domain family, member 1 | |
| Rabbit | |
| Sequential Chromatography | |
| RUO | |
| 23216, 360937, 57915 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q60949, Q86TI0 | |
| TBC1D1 | |
| A synthesized peptide derived from human TBC1D1(Accession Q86TI0), corresponding to amino acid residues around phosphorylated Ser627. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction