missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-Trf2 (Thr188) Polyclonal Antibody
GREENER_CHOICE

Product Code. 16682375
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16682375 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16682375 Supplier Invitrogen™ Supplier No. PA5105937

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of TRF2 only when phosphorylated at Thr188.

TRF2 is a ubiquitously expressed protein that is implicated in the control of telomere length. TRF2, like TRF1 contains a Myb-related DNA binding motif. It binds to duplex TTAGGG repeats and is localized to all human telomeres in metaphase chromosomes. TRF2 is thought to protect chromosome ends by maintaining the correct structure at telomere termini. The use of mutant forms of TRF2 has implicated a role for TRF2 in the prevention of senescence in primary human cells. Recently, it has been shown that inhibition of TRF2 resulted in apoptosis in a subset of mammalian cell types.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-Trf2 (Thr188)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TERF2
Gene Accession No. O35144, Q15554
Gene Alias Telomeric DNA-binding protein; telomeric repeat binding factor 2; telomeric repeat binding protein 2; telomeric repeat-binding factor 2; TERF2; TRBF2; TRF2; TTAGGG repeat-binding factor 2
Gene Symbols TERF2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TERF2(Accession Q15554), corresponding to amino acid residues around phosphorylated Thr188.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21750, 361403, 7014
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.