missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-TRPV1 (Ser502) Polyclonal Antibody

Product Code. 16476159
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476159 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16476159 Supplier Invitrogen™ Supplier No. PA5106215

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of VR1 only when phosphorylated at Ser502.

TRPV1 (Transient Receptor Potential Cation Channel Subfamily V Member 1) is a receptor for capsaicin, the main pungent ingredient in hot chili peppers, and elicits a sensation of burning pain by selectively activating sensory neurons. TRPV1 is a non-selective cation channel that is structurally related to members of the TRP family of ion channels such as TRPC3 and TRPC6. TRPV1 is permeable to calcium, magnesium, and monovalent cations of sodium and potassium. Structurally, TRPV1 contains 6 trans-membrane domains, a P-loop membrane domain and 3 ankyrin repeat domains. TRPV1 is also activated by increases in temperature in the noxious range, suggesting that it functions as a transducer of painful thermal stimuli in vivo. TRPV1 seems to mediate proton influx and may be involved in intracellular acidosis in nociceptive neurons. The expression of TRPV1 appears to be limited to neurons within the sensory ganglia. Four transcript variants encoding TRPV1 with variations in th 5' UTR sequence have been described. Diseases associated with TRPV1 include Irritable Bowel Syndrome (IBS) and Bladder Disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-TRPV1 (Ser502)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TRPV1
Gene Accession No. O35433, Q704Y3, Q8NER1
Gene Alias Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; osm-9-like TRP channel 1; OTRPC1; si:dkey-9e15.2; transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential V1 channel; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1/2; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; Vr1; VR-1; Vr1l; Vr1l1
Gene Symbols TRPV1
Host Species Rabbit
Immunogen A synthesized peptide derived from human TRPV1(Accession Q8NER1), corresponding to amino acid residues around phosphorylated Ser502.
Purification Method Sequential Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 193034, 7442, 83810
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.