missing translation for 'onlineSavingsMsg'
Learn More

Phospho-TSC2 (Ser939) Rabbit Recombinant Antibody, Proteintech

Product Code. 30235126 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235126 20 μL 20µL
30237799 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235126 Supplier Proteintech Supplier No. 816541RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The Phospho-TSC2 (Ser939) antibody from Proteintech is a rabbit recombinant antibody generated with peptide of human Phospho-TSC2 (Ser939). This antibody recognizes human antigen. The Phospho-TSC2 (Ser939) antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Antigen TSC2 (Ser939)
Applications Western Blot, ELISA
Classification Recombinant Monoclonal
Clone 2B18
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols FLJ43106; LAM; TSC4
Host Species Rabbit
Immunogen Peptide
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7249
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 200 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.