Learn More
Invitrogen™ Phospho-TUBA1C (Ser165) Polyclonal Antibody

Description
This antibody was cross-adsorbed to unphosphorylated alpha6-Tubulin (a.a. 160-169) peptide before affinity purification using alpha6-Tubulin (Ser-165) peptide (without carrier). The antibody detects a 50 kDa* protein corresponding to the molecular mass of alpha-Tubulin on SDS-PAGE immunoblots of recombinant a1-tubulin phosphorylated with PKCalpha. This reactivity is not observed after lambda phosphatase treatment.
Specifications
Specifications
| Antigen | Phospho-TUBA1C (Ser165) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | TUBA1C |
| Gene Accession No. | P68373, Q6AYZ1, Q9BQE3 |
| Gene Alias | alpha-tubulin 6; Alpha-tubulin isotype M-alpha-6; bcm948; Detyrosinated tubulin alpha-1C chain; M[a]6; Tuba1c; TUBA6; tubulin alpha 1c; tubulin alpha-1C chain; Tubulin alpha-6 chain; tubulin, alpha 1C; tubulin, alpha 6 |
| Gene Symbols | TUBA1C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.