missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Antibody (PY265), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08299R
This item is not returnable.
View return policy
Description
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence
Specifications
| Phospho-Tyrosine | |
| Monoclonal | |
| DyLight 550 | |
| 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine | |
| Phosphotyrosine conjugated to BSA | |
| 100 μg | |
| Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| Non-species specific | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence | |
| PY265 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido