missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Tyrosine Antibody (PY2870R), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08423APC
This item is not returnable.
View return policy
Description
Phospho-Tyrosine Monoclonal antibody specifically detects Phospho-Tyrosine in Nonspecies specific samples. It is validated for ELISA
Specifications
| Phospho-Tyrosine | |
| Monoclonal | |
| APC | |
| 2 amino 3(4 hydroxyphenyl) propanoic acid, 4 hydroxyphenylalanine, phosphotyrosine, pTyrosine, pY20, Tyrosine | |
| Recombinant full-length human Phospho-Tyrosine protein | |
| 0.1 mL | |
| Neuroscience, Phospho Specific, Signal Transduction, Tyrosine Kinases | |
| Non-species specific | |
| Purified |
| ELISA | |
| PY2870R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction