Learn More
Invitrogen™ Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117247
Description
Cellular location: axon, cytoplasm, cytoplasmic side of plasma membrane, cytoplasmic vesicle, cytosol, dendrite, mitochondrion, neuron projection, nucleus, perikaryon, perinuclear region of cytoplasm, smooth endoplasmic reticulum, synaptic vesicle, terminal bouton Immunogen sequence: RQSLI.
Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.
Specifications
| Phospho-Tyrosine Hydroxylase (Ser40) | |
| Polyclonal | |
| Unconjugated | |
| TH | |
| dystonia 14; DYT14; DYT5b; EC 1.14.16.2; HGNC:11782; Th; TH isoform 3; TH isoform a; th1; TH-4; The; TY3H; TYH; TYH antibody; Tyrosine 3-hydroxylase; tyrosine 3-monooxygenase; tyrosine hydroxylase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25085 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.94 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P04177 | |
| TH | |
| A synthetic phosphorylated peptide around S40 of human Tyrosine Hydroxylase (NP_0003512?). | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.