missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospho-USP28 (Ser67) Polyclonal Antibody
GREENER_CHOICE

Product Code. 15712796
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15712796 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15712796 Supplier Invitrogen™ Supplier No. PA564727

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospho-USP28 (Ser67) Polyclonal Antibody detects endogenous levels of USP28 only when phosphorylated at Ser67.

Modification of target proteins by ubiquitin participates in a wide array of biological functions. Proteins destined for degradation or processing via the 26 S proteasome are coupled to multiple copies of ubiquitin. However, attachment of ubiquitin or ubiquitin-related molecules may also result in changes in subcellular distribution or modification of protein activity. An additional level of ubiquitin regulation, deubiquitination, is catalyzed by proteases called deubiquitinating enzymes, which fall into four distinct families. Ubiquitin C-terminal hydrolases, ubiquitin-specific processing proteases (USPs),1 OTU-domain ubiquitin-aldehyde-binding proteins, and Jab1/Pad1/MPN-domain-containing metallo-enzymes. Among these four families, USPs represent the most widespread and represented deubiquitinating enzymes across evolution. USPs tend to release ubiquitin from a conjugated protein. They display similar catalytic domains containing conserved Cys and His boxes but divergent N-terminal and occasionally C-terminal extensions, which are thought to function in substrate recognition, subcellular localization, and protein-protein interactions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-USP28 (Ser67)
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene USP28
Gene Accession No. Q96RU2
Gene Alias 9830148O20Rik; AU022237; Deubiquitinating enzyme 28; Kiaa1515; mKIAA1515; ubiquitin carboxyl-terminal hydrolase 28; ubiquitin carboxyl-terminal hydrolase 28 variant 1; ubiquitin specific peptidase 28; ubiquitin specific protease 28; ubiquitin thioesterase 28; ubiquitin thiolesterase 28; ubiquitin-specific-processing protease 28; Usp28
Gene Symbols USP28
Host Species Rabbit
Immunogen A synthesized peptide derived from human USP28 (Phospho-Ser67).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57646
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.