missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-VASP (Thr278) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143786
This item is not returnable.
View return policy
Description
The antibody detects a 50 kDa* band corresponding to VASP in Western blots of human A431 and HeLa cells, as well as mouse C2C12 cells that have been treated with calyculin A.
Vasodilator-stimulated phosphoprotein (VASP) is a member of the Ena-VASP protein family. Ena-VASP family members contain an EHV1 N-terminal domain that binds proteins containing E/DFPPPPXD/E motifs and targets Ena-VASP proteins to focal adhesions. In the mid-region of the protein, family members have a proline-rich domain that binds SH3 and WW domain-containing proteins. Their C-terminal EVH2 domain mediates tetramerization and binds both G and F actin. VASP is associated with filamentous actin formation and likely plays a widespread role in cell adhesion and motility. VASP may also be involved in the intracellular signaling pathways that regulate integrin-extracellular matrix interactions. VASP is regulated by the cyclic nucleotide-dependent kinases PKA and PKG.
Specifications
| Phospho-VASP (Thr278) | |
| Polyclonal | |
| Unconjugated | |
| VASP | |
| Enah/Vasp-like; I79_012660; Unknown (protein for MGC:127789); vasodilator stimulated phosphoprotein; vasodilator-stimulated phosphoprotein; vasodilator-stimulated phosphoprotein (VASP); VASP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22323, 361517, 7408 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.25 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| P50552, P70460 | |
| VASP | |
| Phospho-VASP (Thr-278) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding threonine 278 in human VASP. This sequence has significant homology to the conserved site in rat and mouse VASP, but is not conserved in mena or EVL proteins. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction