missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phospho-Vimentin (Ser83) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA517229
This item is not returnable.
View return policy
Description
It is not recommended to aliquot this antibody.
Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
Specifications
| Phospho-Vimentin (Ser83) | |
| Polyclonal | |
| Unconjugated | |
| Vim | |
| cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; HEL113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22352, 7431, 81818 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 37 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| P08670, P20152, P31000 | |
| Vim | |
| Synthetic phosphopeptide corresponding to residues surrounding pSer83 of human vimentin. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction