missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-ZAP70/Syk (Tyr493, Tyr526) Rabbit anti-Human, Mouse, APC, Clone: Zap70Y493-H11, Invitrogen™
Rabbit Monoclonal Antibody
Brand: Thermo Scientific MA536918
This item is not returnable.
View return policy
Description
Phospho-ZAP70/Syk (Tyr493, Tyr526) Monoclonal antibody specifically detects Phospho-ZAP70/Syk (Tyr493, Tyr526) in Human, Mouse samples. It is validated for Flow Cytometry
Specifications
| Phospho-ZAP70/Syk (Tyr493, Tyr526) | |
| Monoclonal | |
| APC | |
| ZAP70 | |
| 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Srk; STCD; STD; syk-related protein tyrosine kinase; Syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; TZK; ZA70; Zap70; ZAP-70; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr493 of human phospho Zap70. | |
| 100 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| Zap70Y493-H11 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P43403, P43404 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 22637, 7535 | |
| 2-8°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction