Learn More
Phospho-ZAP70/Syk (Tyr493, Tyr526) Rabbit anti-Human, Mouse, APC, Clone: Zap70Y493-H11, Invitrogen™
Rabbit Monoclonal Antibody
Brand: Thermo Scientific MA537232
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Phospho-ZAP70/Syk (Tyr493, Tyr526) Monoclonal antibody specifically detects Phospho-ZAP70/Syk (Tyr493, Tyr526) in Human, Mouse samples. It is validated for Flow Cytometry
Tekniske data
| Phospho-ZAP70/Syk (Tyr493, Tyr526) | |
| Monoclonal | |
| APC | |
| ZAP70 | |
| 70 kDa zeta-associated protein; 70 kDa zeta-chain associated protein; ADMIO2; EC 2.7.10.2; FLJ17670; FLJ17679; I79_004087; IMD48; kinase ZAP70; mrtle; mur; Srk; STCD; STD; syk-related protein tyrosine kinase; Syk-related tyrosine kinase; Tyrosine-protein kinase ZAP-70; TZK; ZA70; Zap70; ZAP-70; zeta chain of T cell receptor associated protein kinase 70; zeta chain of T cell receptor associated protein kinase 70kDa; zeta-chain (TCR) associated protein kinase; zeta-chain (TCR) associated protein kinase 70; zeta-chain (TCR) associated protein kinase 70kDa; zeta-chain associated protein kinase 70kDa; zeta-chain associated protein kinase, 70kD | |
| A synthetic phospho-peptide corresponding to residues surrounding Tyr493 of human phospho Zap70. | |
| 10 Tests | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| Zap70Y493-H11 | |
| PBS with 0.20% BSA and 0.09% sodium azide | |
| P43403, P43404 | |
| Rabbit | |
| Protein A/G | |
| RUO | |
| 22637, 7535 | |
| 2-8°C | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion