missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOSPHO2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | PHOSPHO2 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18109167
|
Novus Biologicals
NBP2-38218 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18682476
|
Novus Biologicals
NBP2-38218-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
PHOSPHO2 Polyclonal specifically detects PHOSPHO2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Spezifikation
| PHOSPHO2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| phosphatase, orphan 2 | |
| PHOSPHO2 | |
| IgG | |
| Affinity Purified |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q8TCD6 | |
| 493911 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DSNSVFIDWVLEAASFHDIFDKVFTNPAAFNSNGHLTVENYHTHSCNRCPKNLCKKVVLIEFVDKQLQQGVNYT | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts