missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphodiesterase 4A/PDE4A Antibody (OTI1C8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73376B
This item is not returnable.
View return policy
Description
Phosphodiesterase 4A/PDE4A Monoclonal specifically detects Phosphodiesterase 4A/PDE4A in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Phosphodiesterase 4A/PDE4A | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| PDE4A | |
| Full length recombinant protein of human Phosphodiesterase 4A/PDE4A (NP_006193) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1C8 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| cAMP-specific 3′-5′-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4 | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Phospho Specific, Signal Transduction, Stem Cell Markers | |
| 5141.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction