missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoglucomutase 5 Antibody (14F8/F8), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08656JF646
This item is not returnable.
View return policy
Description
Phosphoglucomutase 5 Monoclonal antibody specifically detects Phosphoglucomutase 5 in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Phosphoglucomutase 5 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| aciculin, EC 5.4.2.2, PGM-RP, PGMRPphosphoglucomutase-like protein 5, phosphoglucomutase 5, Phosphoglucomutase-related protein | |
| Cytoskeletal fraction of the human myometrium tissue extract (Uniprot: Q15124) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 14F8/F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 5239 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction