missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase C beta 2 Antibody, Novus Biologicals™

Product Code. 18602385 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18602385 25 μg 25µL
18655874 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18602385 Supplier Novus Biologicals Supplier No. NBP32132525ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Phospholipase C beta 2 Polyclonal antibody specifically detects Phospholipase C beta 2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospholipase C beta 2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias EC 3.1.4.11, FLJ38135, Phosphoinositide phospholipase C-beta-2,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-2, phospholipase C, beta 2, Phospholipase C-beta-2, PLC-beta-2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: PINALNSGYHHLCLHSESNMPLTMPALFIFLEMKDYIPGAWADLTVALANPIKFFSAHDTKSVKLKEAMGGLPEKPFPLASPVASQVN
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5330
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.