missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipase C like 1 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06275PE
This item is not returnable.
View return policy
Descrizione
Phospholipase C like 1 Polyclonal antibody specifically detects Phospholipase C like 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| Phospholipase C like 1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin | |
| but catalytically inactive protein, inactive phospholipase C-like protein 1, MGC126580, MGC138190, phospholipase C, epsilon, phospholipase C-deleted in lung carcinoma, phospholipase C-like 1, phospholipase C-related but catalytically inactive protein, PLCE, PLCL, PLC-L1, PRIP | |
| Produced in rabbits immunized with E. coli-derived Human Phospholipase C like 1 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 5334 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto