missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipid Scramblase 1/PLSCR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32588
This item is not returnable.
View return policy
Description
Phospholipid Scramblase 1/PLSCR1 Polyclonal specifically detects Phospholipid Scramblase 1/PLSCR1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Phospholipid Scramblase 1/PLSCR1 | |
| Polyclonal | |
| Unconjugated | |
| PBS pH7, 1% BSA, 20% Glycerol with 0.01% Thimerosal | |
| PLSCR1 | |
| Recombinant protein encompassing a sequence within the center region of human Phospholipid scramblase 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 5359 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.52mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1 | |
| Rabbit | |
| 35 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction