missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospholipid Scramblase 1/PLSCR1 Antibody (SR1591), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22122-50ul
This item is not returnable.
View return policy
Description
Phospholipid Scramblase 1/PLSCR1 Monoclonal antibody specifically detects Phospholipid Scramblase 1/PLSCR1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications
| Phospholipid Scramblase 1/PLSCR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| SR1591 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Erythrocyte phospholipid scramblase, MmTRA1b, MMTRA1BCa(2+)-dependent phospholipid scramblase 1, phospholipid scramblase 1, PL scramblase 1 | |
| A synthesized peptide derived from human phospholipid Scramblase 1/PLSCR1 (Uniprot #: O15162) | |
| 50 μg | |
| Cardiovascular Biology, Neuroscience, Signal Transduction | |
| 5359 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction