missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphopantothenate-cysteine ligase Antibody (10D11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59481F
This item is not returnable.
View return policy
Description
Phosphopantothenate-cysteine ligase Monoclonal antibody specifically detects Phosphopantothenate-cysteine ligase in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Phosphopantothenate-cysteine ligase | |
| Monoclonal | |
| FITC | |
| FLJ11838, MGC117357, phosphopantothenoylcysteine synthetase, PPC synthetase | |
| Phosphopantothenate-cysteine ligase (1-311aa) purified from E. coli | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 79717 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 10D11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction