missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoribosyl Pyrophosphate Amidotransferase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 550.00
Specifications
| Antigen | Phosphoribosyl Pyrophosphate Amidotransferase |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 |
| Applications | ELISA, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231264
|
Novus Biologicals
NBP3-38312-20ul |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228565
|
Novus Biologicals
NBP3-38312-100ul |
100 μL |
€ 550.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphoribosyl Pyrophosphate Amidotransferase Polyclonal antibody specifically detects Phosphoribosyl Pyrophosphate Amidotransferase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ImmunofluorescenceSpecifications
| Phosphoribosyl Pyrophosphate Amidotransferase | |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.3), 50% glycerol | |
| 5471 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ATASE, EC 2.4.2.14, glutamine phosphoribosylpyrophosphatate amidotransferase, Glutamine phosphoribosylpyrophosphate amidotransferase, glutamine PRPP amidotransferase, GPATamidophosphoribosyltransferase, phosphoribosyl pyrophosphate amidotransferase, PRAT | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human Phosphoribosyl Pyrophosphate Amidotransferase (NP_002694.3).,, Sequence:, MELEELGIREECGVFGCIASGEWPTQLDVPHVITLGLVGLQHRGQESAGIVTSDGSSVPTFKSHKGMGLVNHVFTEDNLKKLYVSNLGIGHTRYATTGKCELENCQPFVVETLHGKIAVAHNGELVNAARLRKKLLRHGIGLSTSSDSEMITQLLAYTPPQEQDDTPDWVARIKNLMKEAPTAYSLLIMHRDVIYAVRDPYGNRPLCIGRLIPVSDINDKEKKTSETEGWVVSSESCSFLSIGARYYREVLPGEIVEISRHNVQTLDIISRSEGNPVAFC | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title