missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoribosyl Pyrophosphate Amidotransferase Antibody (OTI1B8), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73394AF350
This item is not returnable.
View return policy
Description
Phosphoribosyl Pyrophosphate Amidotransferase Monoclonal specifically detects Phosphoribosyl Pyrophosphate Amidotransferase in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Phosphoribosyl Pyrophosphate Amidotransferase | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PPAT | |
| Recombinant protein fragment corresponding to amino acids 42-278 of human Phosphoribosyl Pyrophosphate Amidotransferase (NP_002694) produced in E.coli. | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1B8 | |
| Western Blot, Immunohistochemistry | |
| ATASE, EC 2.4.2.14, glutamine phosphoribosylpyrophosphatate amidotransferase, Glutamine phosphoribosylpyrophosphate amidotransferase, glutamine PRPP amidotransferase, GPATamidophosphoribosyltransferase, phosphoribosyl pyrophosphate amidotransferase, PRAT | |
| Mouse | |
| 0.1 mL | |
| 5471 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction