missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid Dibenzyl Ester Silver Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Phosphoric Acid Dibenzyl Ester Silver Salt |
| CAS | 50651-75-7 |
| Molecular Formula | C14H14AgO4P |
| Quantity | 250 mg |
| Synonym | Dibenzyl Hydrogen Phosphate Silver Salt, Dibenzyl Phosphate Silver Salt, Dibenzylphosphoric Acid Silver Salt |
| InChI Formula | InChI=1S/C14H15O4P.Ag/c15-19(16,17-11-13-7-3-1-4-8-13)18-12-14-9-5-2-6-10-14;/h1-10H,11-12H2,(H,15,16);/q;+1/p-1 |
| SMILES | [Ag+].[O-]P(=O)(OCc1ccccc1)OCc2ccccc2 |
| IUPAC Name | silver;dibenzyl phosphate |
| Molecular Weight (g/mol) | 385.1 |
| Formula Weight | 383.9681 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?